Learn More
Waters Corp Viridis HSS C18 SB Column, 100Å, 1.8 µm, 3 mm X 75 mm, 1/pk
Catalog No. 50466373 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Viridis sub-2-µm and 3.5 µm columns demonstrate high efficiency and resolution. The reduction and control of surface silanol activity on Viridis particles delivers, under SFC conditions, excellent peak shapes — without the use of mobile-phase additives — even for well-retained basic achiral compounds.
Catalog No. 50-466-373 Supplier Waters Corp Supplier No. 186006622
The Viridis sub-2-µm and 3.5 µm columns demonstrate high efficiency and resolution. The reduction and control of surface silanol activity on Viridis particles delivers, under SFC conditions, excellent peak shapes — without the use of mobile-phase additives — even for well-retained basic achiral compounds.