Learn More
Waters Corp Waters Viridis HSS C18 SB Column, 1.8 μm, 2.1 mm × 75 mm
Catalog No. 50466369 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Viridis sub-2-μm and 3.5 μm columns are designed and tested for the ACQUITY UPC2 system, providing high efficiency and resolution. These columns feature a reduction and control of surface silanol activity on Viridis particles, which ensures excellent peak shapes under SuperCritical Fluid (SFC) conditions, even for well-retained basic achiral compounds, without the need for mobile-phase additives.
  • Designed and tested for the ACQUITY UPC2 system.
  • Demonstrates high efficiency and resolution.
  • Features reduction and control of surface silanol activity on particles.
  • Delivers excellent peak shapes under SFC conditions.
  • Achieves excellent peak shapes without the use of mobile-phase additives.
  • Effective for well-retained basic achiral compounds.
Catalog No. 50-466-369 Supplier Waters Corp Supplier No. 186006618
The Viridis sub-2-μm and 3.5 μm columns are designed and tested for the ACQUITY UPC2 system, providing high efficiency and resolution. These columns feature a reduction and control of surface silanol activity on Viridis particles, which ensures excellent peak shapes under SuperCritical Fluid (SFC) conditions, even for well-retained basic achiral compounds, without the need for mobile-phase additives.
  • Designed and tested for the ACQUITY UPC2 system.
  • Demonstrates high efficiency and resolution.
  • Features reduction and control of surface silanol activity on particles.
  • Delivers excellent peak shapes under SFC conditions.
  • Achieves excellent peak shapes without the use of mobile-phase additives.
  • Effective for well-retained basic achiral compounds.