Learn More
Waters Corp WATERS CORPORATION VanGuard Pre-Column, HSS C18 SB, 1.7 μm, 2.1 × 5 mm
Catalog No. 50466367 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This VanGuard Pre-Column is designed for high-speed applications, offering protection for analytical columns. It features HSS C18 SB chemistry with a 1.7 μm particle size and compact dimensions of 2.1 × 5 mm.
  • VanGuard Pre-Column design.
  • HSS C18 SB chemistry.
  • 1.7 μm particle size.
  • 2.1 × 5 mm dimensions.
  • Supplied in a pack of 3.
Catalog No. 50-466-367 Supplier Waters Corp Supplier No. 186006616
This VanGuard Pre-Column is designed for high-speed applications, offering protection for analytical columns. It features HSS C18 SB chemistry with a 1.7 μm particle size and compact dimensions of 2.1 × 5 mm.
  • VanGuard Pre-Column design.
  • HSS C18 SB chemistry.
  • 1.7 μm particle size.
  • 2.1 × 5 mm dimensions.
  • Supplied in a pack of 3.