Learn More
Waters Corp Test Solutions, UV Detector, 25 mm Cell, Liquid Formulation
Supplier:  Waters Corp 186006694
Encompass_Preferred
This product is a kit of liquid standards designed for use with UV detectors equipped with 25 mm flow cells in UPLC systems. It provides a comprehensive solution for verifying various aspects of system performance.
  • Designed for use on UV detectors with a 25 mm flow cell.
  • Contains standards for verifying system reproducibility.
  • Enables verification of flow rate and gradient performance.
  • Supports testing of detector linearity and accuracy.
  • Facilitates assessment of injector accuracy and carryover.
  • Provided in a liquid formulation for ease of use.
  • Stable at room temperature for convenient storage.
  • Offers a shelf life of 24 months.
  • Each ampoule contains 10 mL of solution.
Catalog No. 50-239-1139