Learn More
Waters Corp Standards, System Performance, UV Detection, 10 mL Ampoule
Catalog No. 502037723 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This ACQUITY UPLC standards kit is designed for comprehensive system assessment. It includes standards suitable for verifying system reproducibility, flow rate, gradient performance, detector linearity, detector accuracy, injector accuracy, and injector carryover.
  • Appropriate for verification of system reproducibility.
  • Appropriate for verification of flow rate.
  • Appropriate for verification of gradient performance.
  • Appropriate for verification of detector linearity.
  • Appropriate for verification of detector accuracy.
  • Appropriate for verification of injector accuracy.
  • Appropriate for verification of injector carryover.
Catalog No. 50-203-7723 Supplier Waters Corp Supplier No. 186006682
This ACQUITY UPLC standards kit is designed for comprehensive system assessment. It includes standards suitable for verifying system reproducibility, flow rate, gradient performance, detector linearity, detector accuracy, injector accuracy, and injector carryover.
  • Appropriate for verification of system reproducibility.
  • Appropriate for verification of flow rate.
  • Appropriate for verification of gradient performance.
  • Appropriate for verification of detector linearity.
  • Appropriate for verification of detector accuracy.
  • Appropriate for verification of injector accuracy.
  • Appropriate for verification of injector carryover.