Learn More
Waters Corp ACQUITY UPLC Absorb Start-up Solution, UV Detection, LC System, 10 mL per Ampoule
Catalog No. 502392776 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Acquity UPLC standards are provided in two 10 mL ampules. Solution 1 contains 0.1000 mg/mL Caffeine ± 0.5%, and Solution 2 contains 4 μg/mL ± 1.0% each of 2-Acetylfuran, Acetanilide, Acetophenone, Butylparaben, Benzophenone, and Valerophenone. This product is specific for sale to China as an alternative to 700002669.
  • UV detection
  • Liquid formulation
  • Room temperature storage
  • LC system type
  • ACQUITY instrument compatible
  • Quantitative analysis capability
  • 24 months shelf life
  • Ambient shipping condition
  • 10 mL volume per ampoule
  • Reference type
  • Designed for system performance application
  • Standards product type
  • One unit per package
Catalog No. 50-239-2776 Supplier Waters Corp Supplier No. 186006683
Acquity UPLC standards are provided in two 10 mL ampules. Solution 1 contains 0.1000 mg/mL Caffeine ± 0.5%, and Solution 2 contains 4 μg/mL ± 1.0% each of 2-Acetylfuran, Acetanilide, Acetophenone, Butylparaben, Benzophenone, and Valerophenone. This product is specific for sale to China as an alternative to 700002669.
  • UV detection
  • Liquid formulation
  • Room temperature storage
  • LC system type
  • ACQUITY instrument compatible
  • Quantitative analysis capability
  • 24 months shelf life
  • Ambient shipping condition
  • 10 mL volume per ampoule
  • Reference type
  • Designed for system performance application
  • Standards product type
  • One unit per package