Learn More
Waters Corp Active PreHeater (APH), MP35N, .004 in ID x 12.5 in, with Gasket - PLUS
Catalog No. NC1632803 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Replacement Active PreHeater (APH), MP35N, .004 in ID x 12.5 in, with Gasket - PLUS. ; This APH is used in H-Class PLUS Bio systems with CM-A.
Catalog No. NC1632803 Supplier Waters Corp Supplier No. 205001754
Replacement Active PreHeater (APH), MP35N, .004 in ID x 12.5 in, with Gasket - PLUS. ; This APH is used in H-Class PLUS Bio systems with CM-A.