Learn More
Waters Corp Waters ACQUITY UPLC BEH Amide Column, 1.7 μm, 3.0 x 30 mm
Supplier:  Waters Corp 186004803
Encompass_Preferred
ACQUITY UPLC BEH Amide columns use a chemically stable, trifunctionally-bonded amide phase for HILIC separations. They retain polar analytes and metabolites, facilitating a wide range of mobile phase pH (2–11) for exceptional retention. These columns enhance retention of polar compounds, increase mass spectrometry response, and are compatible with sample preparation eluates, offering an attractive separation technique.
  • High resolution and speed for carbohydrate analysis (1.7 μm particle).
  • Increased chemical stability allows high pH/temperature use, shortening analysis and improving MS detection.
  • Exceptional column lifetime and assay robustness via BEH particle technology.
  • Not susceptible to Schiff base formation, improving quantitation accuracy.
  • High pH mobile phase compatibility simplifies sample pre-treatment and improves LC/MS sensitivity.
Catalog No. 50-791-200