Learn More
Waters Corp Waters ACQUITY UPLC BEH Amide Column, 130Å, 1.7 μm, 1.0 x 100 mm
Catalog No. 50465909 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
ACQUITY UPLC BEH Amide columns feature a chemically stable, trifunctionally-bonded amide phase with ethylene bridged hybrid (BEH) particle technology. Designed for Hydrophilic Interaction Chromatography (HILIC), these columns effectively retain and separate extremely polar compounds, offering stability and versatility across a wide mobile phase pH range (2–11). They also enhance mass spectrometry response and provide orthogonal selectivity.
  • HILIC separations for retaining and separating extremely polar compounds.
  • Wide mobile phase pH versatility (2–11) for broad analyte retention.
  • Increased mass spectrometry response and direct compatibility with sample preparation eluates.
  • Orthogonal selectivity compared to reversed-phase materials.
  • High-resolution and high-speed carbohydrate analysis with 1.7 μm particle size.
Catalog No. 50-465-909 Supplier Waters Corp Supplier No. 176001915
ACQUITY UPLC BEH Amide columns feature a chemically stable, trifunctionally-bonded amide phase with ethylene bridged hybrid (BEH) particle technology. Designed for Hydrophilic Interaction Chromatography (HILIC), these columns effectively retain and separate extremely polar compounds, offering stability and versatility across a wide mobile phase pH range (2–11). They also enhance mass spectrometry response and provide orthogonal selectivity.
  • HILIC separations for retaining and separating extremely polar compounds.
  • Wide mobile phase pH versatility (2–11) for broad analyte retention.
  • Increased mass spectrometry response and direct compatibility with sample preparation eluates.
  • Orthogonal selectivity compared to reversed-phase materials.
  • High-resolution and high-speed carbohydrate analysis with 1.7 μm particle size.