Learn More
Waters Corp Waters ACQUITY UPLC BEH Amide Column, 1.7 μm, 1 mm × 150 mm, 130 Å
Catalog No. 50465910 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The ACQUITY UPLC BEH Amide Column, utilizing BEH technology and a chemically stable amide phase, is engineered for efficient HILIC separation of highly polar compounds like carbohydrates. It offers high resolution and robust performance across a broad pH range.
  • Retains extremely polar compounds, including carbohydrates and sugars.
  • Utilizes BEH technology with a trifunctionally bonded amide phase.
  • Stable from pH 2 to 11 for broad application.
  • Offers high resolution, high speed analysis with 1.7 μm particle size.
  • Ensures exceptional column lifetime and assay robustness.
  • Optimized for Mass Spectrometry (MS) response.
  • Avoids Schiff base formation, improving carbohydrate quantitation.
  • Compatible with UPLC/UHPLC systems; includes eCord™ technology.
  • Withstands maximum pressure of 18000 psi (1240 Bar).
Catalog No. 50-465-910 Supplier Waters Corp Supplier No. 176001916
The ACQUITY UPLC BEH Amide Column, utilizing BEH technology and a chemically stable amide phase, is engineered for efficient HILIC separation of highly polar compounds like carbohydrates. It offers high resolution and robust performance across a broad pH range.
  • Retains extremely polar compounds, including carbohydrates and sugars.
  • Utilizes BEH technology with a trifunctionally bonded amide phase.
  • Stable from pH 2 to 11 for broad application.
  • Offers high resolution, high speed analysis with 1.7 μm particle size.
  • Ensures exceptional column lifetime and assay robustness.
  • Optimized for Mass Spectrometry (MS) response.
  • Avoids Schiff base formation, improving carbohydrate quantitation.
  • Compatible with UPLC/UHPLC systems; includes eCord™ technology.
  • Withstands maximum pressure of 18000 psi (1240 Bar).