Learn More
Waters Corp Waters Corporation ACQUITY UPLC BEH Amide Column, 130Å, 1.7 μm, 3.0 x 50 mm
Supplier:  Waters Corp 186004804
Encompass_Preferred
These columns are designed for hydrophilic interaction chromatography (HILIC) of extremely polar compounds.
  • Retains extremely polar compounds, including carbohydrates and sugars, in HILIC separations.
  • Utilizes a chemically stable, trifunctionally-bonded amide phase, stable across pH 2 to 11.
  • Offers orthogonal selectivity to reversed-phase materials with increased mass spectrometry response.
  • Features 1.7 μm particle size for high-resolution, high-speed analysis in complex matrices.
  • Enhances assay robustness, column lifetime, and quantitation accuracy by preventing Schiff base formation.
  • High pH mobile phase compatibility simplifies LC/MS analysis and improves sensitivity.
Catalog No. 50-791-201