Learn More
Waters Corp Waters Column, BEH Amide, 130Å, 1.7 μm, 3 mm × 150 mm
Catalog No. 50791202 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The ACQUITY BEH Amide column retains extremely polar compounds, such as carbohydrates and sugars. It incorporates BEH technology with a chemically stable, trifunctionally bonded amide phase, stable from pH 2 to 11. This design supports HILIC separations of polar analytes across diverse polarities and pKa values.
  • Retains polar compounds like carbohydrates and sugars.
  • Chemically stable, trifunctionally bonded amide phase (pH 2 to 11).
  • Efficient, repeatable UPLC HILIC separations.
  • Increased mass spectrometry response.
  • Compatible with sample preparation elutes (PPT, LLE, SPE).
  • Orthogonal selectivity versus reversed-phase materials.
  • 1.7 μm particle size for high-speed, high-resolution analysis.
  • Enhances quantitation accuracy; avoids Schiff base formation.
  • High pH stability reduces sugar anomers, analysis time, and boosts MS detection.
Catalog No. 50-791-202 Supplier Waters Corp Supplier No. 186004806
The ACQUITY BEH Amide column retains extremely polar compounds, such as carbohydrates and sugars. It incorporates BEH technology with a chemically stable, trifunctionally bonded amide phase, stable from pH 2 to 11. This design supports HILIC separations of polar analytes across diverse polarities and pKa values.
  • Retains polar compounds like carbohydrates and sugars.
  • Chemically stable, trifunctionally bonded amide phase (pH 2 to 11).
  • Efficient, repeatable UPLC HILIC separations.
  • Increased mass spectrometry response.
  • Compatible with sample preparation elutes (PPT, LLE, SPE).
  • Orthogonal selectivity versus reversed-phase materials.
  • 1.7 μm particle size for high-speed, high-resolution analysis.
  • Enhances quantitation accuracy; avoids Schiff base formation.
  • High pH stability reduces sugar anomers, analysis time, and boosts MS detection.