Learn More
Waters Corp Waters ACQUITY UPLC BEH Amide Column, 1.7 μm, 130 Å
Supplier:  Waters Corp 176001908
Encompass_Preferred
ACQUITY BEH Amide columns are designed to retain extremely polar compounds such as carbohydrates and sugars. Utilizing BEH technology and a chemically stable, trifunctionally bonded amide phase, these columns maintain stability from pH 2 to 12, enabling versatile separation of polar analytes.
  • Retains extremely polar compounds.
  • Chemically stable, trifunctionally bonded amide phase.
  • Stable from pH 2 to 12.
  • 1.7 μm particle size for high resolution and speed.
  • Superior column lifetime with BEH particle technology.
  • Increased MS response; compatible with sample preparation eluates.
  • Orthogonal selectivity to reversed-phase materials.
Catalog No. 50-203-7303