Learn More
Waters Corp Waters Corporation ACQUITY UPLC BEH Amide Column, 130Å, 1.7 μm, 2.1 x 100 mm
Catalog No. NC9900976 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
ACQUITY BEH Amide columns are designed to retain extremely polar compounds such as carbohydrates and sugars. Utilizing BEH technology and a chemically stable, trifunctionally bonded amide phase, these columns operate from pH 2 to 12, allowing separation of polar analytes across a wide range of polarity and pKa. They also facilitate a wide range of mobile phase pH (2–11) for analytes too polar for reversed-phase chromatography.
  • Retains extremely polar compounds, including carbohydrates and sugars.
  • Chemically stable trifunctionally bonded amide phase stable from pH 2 to 12.
  • Provides orthogonal separation for polar and ionizable compounds.
  • 1.7 μm particle size enables high-speed, high-resolution analysis.
  • Improved quantitation accuracy by preventing Schiff base formation.
  • Exceptional column lifetime and assay robustness.
Catalog No. NC9900976 Supplier Waters Corp Supplier No. 186004801
ACQUITY BEH Amide columns are designed to retain extremely polar compounds such as carbohydrates and sugars. Utilizing BEH technology and a chemically stable, trifunctionally bonded amide phase, these columns operate from pH 2 to 12, allowing separation of polar analytes across a wide range of polarity and pKa. They also facilitate a wide range of mobile phase pH (2–11) for analytes too polar for reversed-phase chromatography.
  • Retains extremely polar compounds, including carbohydrates and sugars.
  • Chemically stable trifunctionally bonded amide phase stable from pH 2 to 12.
  • Provides orthogonal separation for polar and ionizable compounds.
  • 1.7 μm particle size enables high-speed, high-resolution analysis.
  • Improved quantitation accuracy by preventing Schiff base formation.
  • Exceptional column lifetime and assay robustness.