Learn More
Waters Corp ACQUITY UPLC BEH Amide Column, 1.7 μm, 130 Å, 2.1 × 75 mm
Catalog No. 50466182 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The ACQUITY UPLC BEH Amide Column, utilizing BEH (Ethylene Bridged Hybrid) technology, is engineered for robust retention and separation of highly polar compounds like carbohydrates and sugars. Its chemically stable, trifunctionally bonded amide stationary phase maintains stability across a broad pH range from 2 to 11, facilitating efficient separation of diverse polar analytes and offering enhanced performance in chromatographic applications.
  • Retains extremely polar compounds, including carbohydrates and sugars.
  • Features a chemically stable, trifunctionally bonded amide phase.
  • Stable from pH 2 to 11.
  • Enables separation of polar analytes across a wide range of polarity.
  • Provides mass spectrometry response and direct compatibility with sample preparation eluates.
  • Exceptional column lifetime.
  • Not vulnerable to Schiff-based formation for improved quantitation accuracy.
Catalog No. 50-466-182 Supplier Waters Corp Supplier No. 186005657
The ACQUITY UPLC BEH Amide Column, utilizing BEH (Ethylene Bridged Hybrid) technology, is engineered for robust retention and separation of highly polar compounds like carbohydrates and sugars. Its chemically stable, trifunctionally bonded amide stationary phase maintains stability across a broad pH range from 2 to 11, facilitating efficient separation of diverse polar analytes and offering enhanced performance in chromatographic applications.
  • Retains extremely polar compounds, including carbohydrates and sugars.
  • Features a chemically stable, trifunctionally bonded amide phase.
  • Stable from pH 2 to 11.
  • Enables separation of polar analytes across a wide range of polarity.
  • Provides mass spectrometry response and direct compatibility with sample preparation eluates.
  • Exceptional column lifetime.
  • Not vulnerable to Schiff-based formation for improved quantitation accuracy.