Learn More
Waters Corp Waters ACQUITY UPLC BEH Amide Column, 1.7 μm, 3 mm x 150 mm
Catalog No. 50791072 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
  • Retains extremely polar compounds, including carbohydrates and sugars.
  • Features a chemically stable, trifunctionally bonded amide phase based on BEH technology, stable from pH 2 to 12.
  • Enables efficient, reproducible UPLC HILIC separations for a wide range of polar analytes.
  • Provides increased mass spectrometry response and direct compatibility with sample preparation eluates.
  • Offers orthogonal selectivity to reversed-phase materials.
  • 1.7 μm particle size for high-speed, high-resolution analysis.
  • Improves quantitation accuracy by resisting Schiff base formation.
  • Increased pH stability allows for shorter analysis times and improved MS detection.
  • Offers exceptional column lifetime and improved assay robustness.
Catalog No. 50-791-072 Supplier Waters Corp Supplier No. 176001913
  • Retains extremely polar compounds, including carbohydrates and sugars.
  • Features a chemically stable, trifunctionally bonded amide phase based on BEH technology, stable from pH 2 to 12.
  • Enables efficient, reproducible UPLC HILIC separations for a wide range of polar analytes.
  • Provides increased mass spectrometry response and direct compatibility with sample preparation eluates.
  • Offers orthogonal selectivity to reversed-phase materials.
  • 1.7 μm particle size for high-speed, high-resolution analysis.
  • Improves quantitation accuracy by resisting Schiff base formation.
  • Increased pH stability allows for shorter analysis times and improved MS detection.
  • Offers exceptional column lifetime and improved assay robustness.