Learn More
Waters Corp Waters ACQUITY UPLC BEH Amide Column, Amide, 1.7 μm, 1 mm, 150 mm
Supplier:  Waters Corp 186004850
Encompass_Preferred
ACQUITY UPLC BEH Amide columns are designed for Hydrophilic Interaction Chromatography (HILIC). They retain polar compounds like carbohydrates and sugars, difficult for reversed-phase chromatography. With BEH technology and a chemically stable, trifunctionally bonded amide phase, these columns perform robustly across a pH range of 2 to 12 for versatile analyte separation.
  • Retains extremely polar compounds (carbohydrates, sugars) in HILIC separations.
  • Chemically stable, trifunctionally bonded amide phase with BEH technology.
  • Wide pH stability (2 to 12) for broad analyte separation.
  • Increases UPLC analysis speed, sensitivity, and resolution.
  • Offers superior separation speed and sensitivity; withstands pressures up to 18,000 psi (1240 Bar).
  • Compatible with high pH mobile phases for enhanced MS detection.
Catalog No. 50-466-118