Learn More
Waters Corp Waters ACQUITY UPLC BEH Amide Method Validation Kit, 130Å, 1.7 μm, 2.1 mm x 100 mm
Catalog No. 50791204 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The ACQUITY UPLC BEH Amide Method Validation Kit includes three ACQUITY UPLC BEH Amide columns, each packed with a different sorbent batch. These columns feature trifunctionally-bonded amide phase on ethylene bridged hybrid (BEH) particle technology, which supports a wide mobile phase pH range (2–11) and provides exceptional retention for polar analytes, while also increasing mass spectrometry response.
  • Wide mobile phase pH range (2–11) allows for versatile applications.
  • Increased mass spectrometry response and direct compatibility with sample preparation eluates.
  • Offers orthogonal selectivity compared to reversed-phase materials.
  • Designed for exceptional column lifetime, improving assay robustness.
  • 1.7 μm particle size enables high-resolution, high-speed analysis.
  • Not susceptible to Schiff base formation, which improves quantitation accuracy.
Catalog No. 50-791-204 Supplier Waters Corp Supplier No. 186004808
Add to Cart
Add to Cart
The ACQUITY UPLC BEH Amide Method Validation Kit includes three ACQUITY UPLC BEH Amide columns, each packed with a different sorbent batch. These columns feature trifunctionally-bonded amide phase on ethylene bridged hybrid (BEH) particle technology, which supports a wide mobile phase pH range (2–11) and provides exceptional retention for polar analytes, while also increasing mass spectrometry response.
  • Wide mobile phase pH range (2–11) allows for versatile applications.
  • Increased mass spectrometry response and direct compatibility with sample preparation eluates.
  • Offers orthogonal selectivity compared to reversed-phase materials.
  • Designed for exceptional column lifetime, improving assay robustness.
  • 1.7 μm particle size enables high-resolution, high-speed analysis.
  • Not susceptible to Schiff base formation, which improves quantitation accuracy.