Learn More
Waters Corp Waters ACQUITY UPLC BEH C18 Method Validation Kit
Supplier:  Waters Corp 186006019
Encompass_Preferred
This kit consists of three ACQUITY UPLC BEH C18 columns, each packed with a different batch of sorbent. These columns are engineered to endure high pressures and feature efficient 1.7 μm Bridged Ethyl Hybrid (BEH) particles. They enable high speed, sensitivity, and resolution with an optimal blend of ultra-performance efficiencies, a broad pH range, and complementary selectivities.
  • Kit includes three columns, each with a different sorbent batch.
  • Engineered for high-pressure UPLC Technology.
  • Features efficient 1.7 μm Bridged Ethyl Hybrid (BEH) particles.
  • Provides high speed, sensitivity, and resolution.
  • Offers a broad pH range and complementary selectivities.
  • Manufactured in a cGMP, ISO 9001 certified facility.
  • Each column includes a Performance Chromatogram and Certificate of Batch Analysis.
Catalog No. 50-201-505