Learn More
Waters Corp Waters Corporation ACQUITY UPLC BEH C18 Column, 130Å, 1.7 μm, 1x50 mm
Supplier:  Waters Corp 176000861
Encompass_Preferred
Waters Corporation ACQUITY UPLC BEH C18 Column, 130Å, 1.7 μm, 1x50 mm
Catalog No. 50-780-001