Learn More
Waters Corp Waters ACQUITY UPLC BEH C18 Column, 130Å, 1.7 μm, 2.1 x 100 mm
Catalog No. 50780004 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The ACQUITY UPLC BEH C18 column is a premier choice for UPLC method development, featuring a 1.7 μm trifunctionally bonded BEH particle. This design ensures a wide usable pH range of 1 to 12, superior low pH stability, and ultra-low column bleed, making it ideal for high-sensitivity MS applications. It provides exceptional stability for diverse analytes, improving retention, selectivity, and sensitivity with mobile-phase pH.
  • Features 1.7 μm trifunctionally bonded BEH particles.
  • Provides a wide usable pH range (1 to 12) and superior low pH stability.
  • Offers ultra-low column bleed, ideal for high-sensitivity MS applications.
  • Optimized 130Å pore size and 1.7 μm particle size for high-resolution.
  • Enhances separation efficiency, sensitivity, and throughput in UPLC.
  • BEH technology ensures mechanical strength, chemical stability, and extended column lifetime.
Catalog No. 50-780-004 Supplier Waters Corp Supplier No. 176000864
The ACQUITY UPLC BEH C18 column is a premier choice for UPLC method development, featuring a 1.7 μm trifunctionally bonded BEH particle. This design ensures a wide usable pH range of 1 to 12, superior low pH stability, and ultra-low column bleed, making it ideal for high-sensitivity MS applications. It provides exceptional stability for diverse analytes, improving retention, selectivity, and sensitivity with mobile-phase pH.
  • Features 1.7 μm trifunctionally bonded BEH particles.
  • Provides a wide usable pH range (1 to 12) and superior low pH stability.
  • Offers ultra-low column bleed, ideal for high-sensitivity MS applications.
  • Optimized 130Å pore size and 1.7 μm particle size for high-resolution.
  • Enhances separation efficiency, sensitivity, and throughput in UPLC.
  • BEH technology ensures mechanical strength, chemical stability, and extended column lifetime.