Learn More
Waters Corp Waters ACQUITY UPLC BEH C18 Column, 130Å, 1.7 μm, 2.1 mm x 150 mm
Catalog No. 50780023 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This ACQUITY UPLC BEH C18 Column features a 1.7 μm BEH C18 sorbent with trifunctionally bonded particles, ensuring a wide usable pH range of 1–12 and superior low pH stability. Ideal for high sensitivity MS applications, it is designed for up to 15,000 psi (1000 bar) to provide enhanced resolution, sample throughput, and sensitivity for UPLC separations.
  • Universal column for UPLC method development.
  • Features wide usable pH range (1–12) and superior low pH stability.
  • Provides unprecedented efficiency, peak asymmetry, and chemical stability.
  • Designed for applications up to 15,000 psi (1000 bar).
  • Improves resolution, sample throughput, and sensitivity.
  • Offers superior mechanical strength, high pH stability, and excellent peak shape for bases.
Catalog No. 50-780-023 Supplier Waters Corp Supplier No. 176001048
This ACQUITY UPLC BEH C18 Column features a 1.7 μm BEH C18 sorbent with trifunctionally bonded particles, ensuring a wide usable pH range of 1–12 and superior low pH stability. Ideal for high sensitivity MS applications, it is designed for up to 15,000 psi (1000 bar) to provide enhanced resolution, sample throughput, and sensitivity for UPLC separations.
  • Universal column for UPLC method development.
  • Features wide usable pH range (1–12) and superior low pH stability.
  • Provides unprecedented efficiency, peak asymmetry, and chemical stability.
  • Designed for applications up to 15,000 psi (1000 bar).
  • Improves resolution, sample throughput, and sensitivity.
  • Offers superior mechanical strength, high pH stability, and excellent peak shape for bases.