Learn More
Waters Corp ACQUITY UPLC Column, BEH C18, 130Å, 1.7 μm, 2.1 mm × 150 mm
Supplier:  Waters Corp 186002353
Encompass_Preferred
This column is designed for UPLC separations and method development, featuring a trifunctionally bonded BEH particle with a wide usable pH range of 1 to 12. It offers superior low pH stability and ultra-low column bleed, making it ideal for high-sensitivity MS applications. It leverages Ethylene Bridged Hybrid particles for superior peak asymmetry, efficiency, and chemical stability, allowing reliable operation at high backpressure.
  • Universal for UPLC separations and method development.
  • Wide usable pH range (1 to 12) with superior low pH stability.
  • Ultra-low column bleed for high-sensitivity MS applications.
  • Ethylene Bridged Hybrid (BEH) particles provide superior peak asymmetry and chemical stability.
  • 130 Å pore size is versatile for both large and small molecule analysis.
Catalog No. 50-781-086