Learn More
Waters Corp Waters ACQUITY UPLC BEH C18 Column, 1.7 μm, 130 Å, 1 mm x 150 mm
Catalog No. 50781082 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The 1.7 μm ACQUITY UPLC BEH C18 sorbent is designed as a premier column for method development and offers a universal choice for UPLC separations. It features a trifunctionally bonded BEH particle that provides a wide usable pH range (pH 1 to 12), superior stability at low pH, and ultra-low column bleed, making it suitable for high-sensitivity MS applications.
  • Most universal column choice for UPLC separations.
  • Widest usable pH range (pH 1 to 12).
  • Superior low pH stability.
  • Ultra-low column bleed for high-sensitivity MS applications.
  • Engineered to operate effectively over a wide pH range of 1 to 12.
  • Can withstand high pressures up to 18000 psi (1240 bar) without loss of performance or durability.
Catalog No. 50-781-082 Supplier Waters Corp Supplier No. 186002347
The 1.7 μm ACQUITY UPLC BEH C18 sorbent is designed as a premier column for method development and offers a universal choice for UPLC separations. It features a trifunctionally bonded BEH particle that provides a wide usable pH range (pH 1 to 12), superior stability at low pH, and ultra-low column bleed, making it suitable for high-sensitivity MS applications.
  • Most universal column choice for UPLC separations.
  • Widest usable pH range (pH 1 to 12).
  • Superior low pH stability.
  • Ultra-low column bleed for high-sensitivity MS applications.
  • Engineered to operate effectively over a wide pH range of 1 to 12.
  • Can withstand high pressures up to 18000 psi (1240 bar) without loss of performance or durability.