Learn More
Waters Corp Waters ACQUITY UPLC BEH C18 Column, 130Å, 1.7 μm, 1 × 100 mm
Catalog No. 50781081 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The ACQUITY UPLC BEH C18 Column is the premier column for method development, offering a universal choice for UPLC separations. It features a trifunctionally bonded BEH particle, which provides a wide usable pH range (pH 1 to 12), superior low pH stability, and ultra-low column bleed, making it suitable for high sensitivity MS applications. This column helps achieve unprecedented levels of efficiency, peak symmetry, and chemical stability, delivering improved resolution, sensitivity, and speed of analysis.
  • Exceptional low and high-pH stability for all analyte types.
  • High-temperature stability (80 °C).
  • Optimized packing materials for UPLC systems.
  • Manufactured in a cGMP, ISO 9001-certified plant using ultra-pure reagents.
  • Each batch is chromatographically tested with acidic, basic, and neutral analytes.
  • Includes a performance chromatogram and a certificate of batch analysis.
Catalog No. 50-781-081 Supplier Waters Corp Supplier No. 186002346
The ACQUITY UPLC BEH C18 Column is the premier column for method development, offering a universal choice for UPLC separations. It features a trifunctionally bonded BEH particle, which provides a wide usable pH range (pH 1 to 12), superior low pH stability, and ultra-low column bleed, making it suitable for high sensitivity MS applications. This column helps achieve unprecedented levels of efficiency, peak symmetry, and chemical stability, delivering improved resolution, sensitivity, and speed of analysis.
  • Exceptional low and high-pH stability for all analyte types.
  • High-temperature stability (80 °C).
  • Optimized packing materials for UPLC systems.
  • Manufactured in a cGMP, ISO 9001-certified plant using ultra-pure reagents.
  • Each batch is chromatographically tested with acidic, basic, and neutral analytes.
  • Includes a performance chromatogram and a certificate of batch analysis.