Learn More
Waters Corp Waters ACQUITY UPLC Column, BEH C18, 130Å, 1.7 μm, 2.1 mm x 75 mm
Catalog No. 50466140 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This 1.7 μm ACQUITY UPLC BEH C18 sorbent column offers a universal choice for UPLC separations. It features a trifunctionally bonded BEH particle, providing a wide usable pH range (1–12), superior low pH stability, ultra-low column bleed, and high-temperature stability (80 °C) for high sensitivity MS applications.
  • Achieve unprecedented levels of efficiency, peak symmetry, and chemical stability.
  • Utilize the tri-functionally bonded analytical column as a universal method development tool.
  • Leverage a wide mobile phase pH range (1–12) and high-temperature stability (80 °C).
  • Rely on lab equipment manufactured in a cGMP, ISO 9001 certified plant.
  • Ensure excellent, reproducible performance as each batch is chromatographically tested.
  • Tolerates operating pressures up to 18000 psi (1241 bar or 124 MPa).
Catalog No. 50-466-140 Supplier Waters Corp Supplier No. 186005604
This 1.7 μm ACQUITY UPLC BEH C18 sorbent column offers a universal choice for UPLC separations. It features a trifunctionally bonded BEH particle, providing a wide usable pH range (1–12), superior low pH stability, ultra-low column bleed, and high-temperature stability (80 °C) for high sensitivity MS applications.
  • Achieve unprecedented levels of efficiency, peak symmetry, and chemical stability.
  • Utilize the tri-functionally bonded analytical column as a universal method development tool.
  • Leverage a wide mobile phase pH range (1–12) and high-temperature stability (80 °C).
  • Rely on lab equipment manufactured in a cGMP, ISO 9001 certified plant.
  • Ensure excellent, reproducible performance as each batch is chromatographically tested.
  • Tolerates operating pressures up to 18000 psi (1241 bar or 124 MPa).