Learn More
Waters Corp Waters UPLC BEH C18 Column, 1.7 μm, 3 mm × 75 mm
Supplier:  Waters Corp 186005609
Encompass_Preferred
As the premier column for method development, the 1.7 μm BEH C18 sorbent provides the most universal column choice for UPLC separations. The trifunctionally bonded BEH particle gives the widest usable pH range (pH 1 to 12), superior low pH stability, and ultra-low column bleed for high sensitivity MS applications.
  • Premier column for method development
  • Universal column choice for UPLC separations
  • Trifunctionally bonded BEH particle
  • Widest usable pH range (pH 1 to 12)
  • Superior low pH stability
  • Ultra-low column bleed for high sensitivity MS applications
Catalog No. 50-466-145