Learn More
Waters Corp WATERS CORPORATION Column, C18, 1.7 μm, 3 mm × 150 mm
Catalog No. 50791030 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This column is a premier choice for UPLC separations, designed to enhance analysis resolution, sensitivity, and speed. It features trifunctionally bonded BEH particles, offering a wide usable pH range of 1 to 12, superior low pH stability, and ultra-low column bleed, ideal for high-sensitivity MS applications.
  • Designed for high-speed UPLC applications.
  • Features trifunctionally bonded BEH particles.
  • Offers a wide usable pH range of 1 to 12.
  • Provides superior low pH stability and ultra-low column bleed.
  • Suitable for evaluating a diverse range of analytes.
  • Improves analysis resolution, sensitivity, and speed.
  • Achieves ultra-fast analysis and enhanced throughput.
  • Operates safely at pressures up to 18,000 psi (1240 Bar).
  • Equipped with eCord technology.
Catalog No. 50-791-030 Supplier Waters Corp Supplier No. 176001797
Add to Cart
Add to Cart
This column is a premier choice for UPLC separations, designed to enhance analysis resolution, sensitivity, and speed. It features trifunctionally bonded BEH particles, offering a wide usable pH range of 1 to 12, superior low pH stability, and ultra-low column bleed, ideal for high-sensitivity MS applications.
  • Designed for high-speed UPLC applications.
  • Features trifunctionally bonded BEH particles.
  • Offers a wide usable pH range of 1 to 12.
  • Provides superior low pH stability and ultra-low column bleed.
  • Suitable for evaluating a diverse range of analytes.
  • Improves analysis resolution, sensitivity, and speed.
  • Achieves ultra-fast analysis and enhanced throughput.
  • Operates safely at pressures up to 18,000 psi (1240 Bar).
  • Equipped with eCord technology.