Learn More
Waters Corp ACQUITY UPLC Column, C18, 1.7 μm, 3 mm, 30 mm, 130 Å
Catalog No. 50791171 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This 1.7 μm sorbent serves as a premier column for method development, providing a universal choice for UPLC separations. Its trifunctionally bonded particle offers a wide usable pH range (1-12), superior low pH stability, and ultra-low column bleed for high sensitivity MS applications.
  • Trifunctionally bonded hybrid particle.
  • Wide usable pH range from 1 to 12.
  • Superior low and high pH stability for various analytes.
  • Ultra-low column bleed for high sensitivity MS applications.
  • Mobile-phase pH and temperature compatibility up to 80 °C.
  • Exceptional efficiency, peak asymmetry, and chemical stability.
  • Impacts retention, selectivity, and sensitivity of ionizable compounds.
Catalog No. 50-791-171 Supplier Waters Corp Supplier No. 186004659
This 1.7 μm sorbent serves as a premier column for method development, providing a universal choice for UPLC separations. Its trifunctionally bonded particle offers a wide usable pH range (1-12), superior low pH stability, and ultra-low column bleed for high sensitivity MS applications.
  • Trifunctionally bonded hybrid particle.
  • Wide usable pH range from 1 to 12.
  • Superior low and high pH stability for various analytes.
  • Ultra-low column bleed for high sensitivity MS applications.
  • Mobile-phase pH and temperature compatibility up to 80 °C.
  • Exceptional efficiency, peak asymmetry, and chemical stability.
  • Impacts retention, selectivity, and sensitivity of ionizable compounds.