Learn More
Waters Corp ACQUITY UPLC BEH C18 Column, 130Å, 1.7 µm, 3 mm X 100 mm, 3/pk
Catalog No. 50791029 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
As the premier column for method development, the 1.7 um ACQUITY UPLC BEH C18 sorbent provides you with the most universal column choice for UPLC separations. The trifunctionally bonded BEH particle gives you the widest usable pH range (pH 1-12), superior low pH stability, and ultra-low column bleed for high sensitivity MS applications.
Catalog No. 50-791-029 Supplier Waters Corp Supplier No. 176001796
As the premier column for method development, the 1.7 um ACQUITY UPLC BEH C18 sorbent provides you with the most universal column choice for UPLC separations. The trifunctionally bonded BEH particle gives you the widest usable pH range (pH 1-12), superior low pH stability, and ultra-low column bleed for high sensitivity MS applications.