Learn More
Waters Corp ACQUITY UPLC BEH C18 Method Validation Kit, 130Å, 1.7 µm, 3 mm X 100 mm, 3/pk
Catalog No. 50466026 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This kit consists of three ACQUITY UPLC BEH C18 columns packed with three different batches of sorbent.
Catalog No. 50-466-026 Supplier Waters Corp Supplier No. 186004692
This kit consists of three ACQUITY UPLC BEH C18 columns packed with three different batches of sorbent.