Learn More
Waters Corp Waters Column, C4, 1.7 μm, 300 Å, 1 mm x 100 mm
Catalog No. NC0141995 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This column is optimized for protein separation by size, hydrophobicity, and isoelectric point. It offers high efficiency and recovery with stability across a wide pH range and temperatures.
  • QC tested for protein separation.
  • C4 ligand minimizes protein carryover and improves peak capacity.
  • Separates large or hydrophobic biological macromolecules effectively.
  • Provides high efficiency and good recovery.
  • Stable across high and low pH levels and temperatures.
  • Manufactured in a cGMP, ISO9002 certified facility.
  • Batch validated for reproducible performance.
  • Includes a performance test chromatogram and certification of acceptance.
  • 1.7 μm particles for enhanced resolution, sensitivity, and speed.
  • Operates with low system pressure.
  • MaxPeak™ High Performance Surfaces improve analyte recovery and reproducibility.
Catalog No. NC0141995 Supplier Waters Corp Supplier No. 186005590
This column is optimized for protein separation by size, hydrophobicity, and isoelectric point. It offers high efficiency and recovery with stability across a wide pH range and temperatures.
  • QC tested for protein separation.
  • C4 ligand minimizes protein carryover and improves peak capacity.
  • Separates large or hydrophobic biological macromolecules effectively.
  • Provides high efficiency and good recovery.
  • Stable across high and low pH levels and temperatures.
  • Manufactured in a cGMP, ISO9002 certified facility.
  • Batch validated for reproducible performance.
  • Includes a performance test chromatogram and certification of acceptance.
  • 1.7 μm particles for enhanced resolution, sensitivity, and speed.
  • Operates with low system pressure.
  • MaxPeak™ High Performance Surfaces improve analyte recovery and reproducibility.