Learn More
Waters Corp ACQUITY UPLC BEH C8 Method Validation Kit, 130Å, 1.7 µm, 2.1 mm X 150 mm, 3/pk
Supplier:  Waters Corp 186006020
Encompass_Preferred
This kit consists of three ACQUITY UPLC BEH C8 columns packed with three different batches of sorbent.
Catalog No. 50-201-506