Learn More
Waters Corp Waters ACQUITY UPLC BEH C8 Column, 1.0 x 100 mm, 1.7 μm
Supplier:  Waters Corp 176000883
Encompass_Preferred
ACQUITY UPLC™ BEH C8 columns are universal for UPLC™ separations with a wide pH range. They use trifunctional ligand bonding chemistries for superior low pH stability and ultra-low column bleed, combined with high pH stability from 1.7 μm BEH particles. Proprietary endcapping processes ensure outstanding peak shape for bases, leading to sharp peaks, high efficiencies, and maximum MS sensitivities.
  • Designed for UPLC™ separations.
  • Wide pH range (1–12).
  • Trifunctional C8 ligand.
  • Superior low pH stability and ultra-low column bleed.
  • High pH stability due to 1.7 μm BEH particle.
  • Proprietary endcapping for outstanding peak shape for bases.
  • Produces sharp peaks, high efficiencies, and maximum MS sensitivities.
  • Enabled by BEH Technology™ for improved efficiency, strength, and pH range.
Catalog No. 50-780-010