Learn More
Waters Corp Waters ACQUITY UPLC BEH C8 Column, 130 Å, 1.7 μm, 2.1 mm x 30 mm
Supplier:  Waters Corp 186003910
Encompass_Preferred
The ACQUITY BEH C8 sorbent, suitable for UPLC separations, offers enhanced speed, peak shape, and chromatographic performance. Its trifunctionally bonded BEH particles provide a wide usable pH range (1–12), superior low pH stability, and ultra-low column bleed, facilitating faster retention of nonpolar substances.
  • Wide usable pH range: 1–12
  • Superior low pH stability; ultra-low column bleed
  • Faster retention of nonpolar substances
  • Trifunctional ligand bonding ensures low pH stability
  • Extended pH operating range via unique chemistries
  • Special end-capping for superior peak form
  • Achieves high efficiencies, MS sensitivity, sharp peak shapes
  • Manufactured in cGMP, ISO 9001 accredited facility
  • Each batch tested chromatographically for consistent performance
  • Analyzed on eCord™ Intelligent Chip
Catalog No. 50-782-063