Learn More
Waters Corp Waters ACQUITY UPLC BEH C8 Column, 130Å, 1.7 μm, 1 mm X 100 mm
Catalog No. 50781417 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The ACQUITY UPLC BEH C8 Column, featuring trifunctionally bonded BEH particles, offers improved peak shape and chromatographic performance. Designed for faster separations with less retention than C18 sorbents, it provides a wide usable pH range of 1 to 12, superior low pH stability, and ultra-low column bleed.
  • Achieves high efficiencies, MS sensitivity, and sharpest peak shapes.
  • Features hybrid particle substrate and reversed-phase separation mode.
  • Manufactured in a cGMP, ISO 9001 accredited facility.
  • Maximum pressure tolerance up to 18000 psi (1240 Bar).
  • Compatible with UPLC and UHPLC systems.
  • Each column includes an eCord™ Intelligent Chip with Performance Chromatogram and Certificate of Batch Analysis.
Catalog No. 50-781-417 Supplier Waters Corp Supplier No. 186002876
The ACQUITY UPLC BEH C8 Column, featuring trifunctionally bonded BEH particles, offers improved peak shape and chromatographic performance. Designed for faster separations with less retention than C18 sorbents, it provides a wide usable pH range of 1 to 12, superior low pH stability, and ultra-low column bleed.
  • Achieves high efficiencies, MS sensitivity, and sharpest peak shapes.
  • Features hybrid particle substrate and reversed-phase separation mode.
  • Manufactured in a cGMP, ISO 9001 accredited facility.
  • Maximum pressure tolerance up to 18000 psi (1240 Bar).
  • Compatible with UPLC and UHPLC systems.
  • Each column includes an eCord™ Intelligent Chip with Performance Chromatogram and Certificate of Batch Analysis.