Learn More
Waters Corp ACQUITY UPLC BEH C8 Column, 1.7 μm, 1.0 x 150 mm
Catalog No. 50780021 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This UPLC column features Ethylene Bridged Hybrid (BEH) particle technology. It is suitable for reversed-phase and hydrophilic-interaction chromatography and can be used for analyses ranging from small molecules to large biopharmaceuticals. The hybrid particle technology provides intrinsic chemical stability, enabling a wider usable pH range.
  • BEH C8 chemistry
  • 1.7 μm particle size
  • 1.0 x 150 mm dimensions
  • Ethylene Bridged Hybrid (BEH) particle technology
  • Suitable for reversed-phase and hydrophilic-interaction chromatography
  • Applicable for small molecule to large biopharmaceutical analysis
  • Intrinsic chemical stability
  • Wider usable pH range (1–12)
Catalog No. 50-780-021 Supplier Waters Corp Supplier No. 176001046
This UPLC column features Ethylene Bridged Hybrid (BEH) particle technology. It is suitable for reversed-phase and hydrophilic-interaction chromatography and can be used for analyses ranging from small molecules to large biopharmaceuticals. The hybrid particle technology provides intrinsic chemical stability, enabling a wider usable pH range.
  • BEH C8 chemistry
  • 1.7 μm particle size
  • 1.0 x 150 mm dimensions
  • Ethylene Bridged Hybrid (BEH) particle technology
  • Suitable for reversed-phase and hydrophilic-interaction chromatography
  • Applicable for small molecule to large biopharmaceutical analysis
  • Intrinsic chemical stability
  • Wider usable pH range (1–12)