Learn More
Waters Corp Waters Corporation Column, C8, 130 Å, 1.7 μm, 2.1 mm × 50 mm
Catalog No. 50781418 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This C8 sorbent is less retentive than C18 sorbent, designed for increased speed in method development and improved peak shape and chromatographic performance. Its trifunctionally bonded particles offer a wide usable pH range (1 to 12), excellent low pH stability, and minimal column bleed. It suits analyses requiring shorter retention times and faster retention of nonpolar compounds.
  • Less retentive than C18 sorbent for faster method development
  • Improved peak shape and chromatographic performance
  • Trifunctionally bonded particles with usable pH range of 1 to 12
  • Superior low pH stability and ultra-low column bleed
  • Exceptional peak shape for bases via exclusive end-capping processes
  • High efficiencies and maximum MS sensitivities
  • eCord intelligent chip records column performance and history
Catalog No. 50-781-418 Supplier Waters Corp Supplier No. 186002877
This C8 sorbent is less retentive than C18 sorbent, designed for increased speed in method development and improved peak shape and chromatographic performance. Its trifunctionally bonded particles offer a wide usable pH range (1 to 12), excellent low pH stability, and minimal column bleed. It suits analyses requiring shorter retention times and faster retention of nonpolar compounds.
  • Less retentive than C18 sorbent for faster method development
  • Improved peak shape and chromatographic performance
  • Trifunctionally bonded particles with usable pH range of 1 to 12
  • Superior low pH stability and ultra-low column bleed
  • Exceptional peak shape for bases via exclusive end-capping processes
  • High efficiencies and maximum MS sensitivities
  • eCord intelligent chip records column performance and history