Learn More
Waters Corp ACQUITY UPLC BEH C8 Column, 130Å, 1.7 μm, 1 x 50 mm
Catalog No. 50781416 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The ACQUITY BEH C8 sorbent is less retentive compared to the ACQUITY BEH C18 sorbent. A method developer may prefer the speed offered by a less retentive column, while improving peak shape and chromatographic performance. The trifunctionally bonded BEH particles offer the widest usable pH range (1 to 12), superior low pH stability, and ultra-low column bleed.
  • Less retentive compared to C18 sorbent
  • Offers speed for method developers
  • Improves peak shape and chromatographic performance
  • Trifunctionally bonded BEH particles
  • Widest usable pH range (1 to 12)
  • Superior low pH stability
  • Ultra-low column bleed
Catalog No. 50-781-416 Supplier Waters Corp Supplier No. 186002875
Add to Cart
Add to Cart
The ACQUITY BEH C8 sorbent is less retentive compared to the ACQUITY BEH C18 sorbent. A method developer may prefer the speed offered by a less retentive column, while improving peak shape and chromatographic performance. The trifunctionally bonded BEH particles offer the widest usable pH range (1 to 12), superior low pH stability, and ultra-low column bleed.
  • Less retentive compared to C18 sorbent
  • Offers speed for method developers
  • Improves peak shape and chromatographic performance
  • Trifunctionally bonded BEH particles
  • Widest usable pH range (1 to 12)
  • Superior low pH stability
  • Ultra-low column bleed