Learn More
Waters Corp ACQUITY UPLC BEH C8 Column, 130Å, 1.7 µm, 3 mm X 75 mm, 1/pk
Supplier:  Waters Corp 186005611
Encompass_Preferred
The ACQUITY BEH C8 sorbent is less retentive compared to the ACQUITY BEH C18 sorbent. A method developer may prefer the speed offered by a less retentive column, while improving peak shape and chromatographic performance. The trifunctionally bonded BEH particles offer the widest usable pH range (1-12), superior low pH stability, and ultra-low column bleed.
Catalog No. 50-466-147