Learn More
Waters Corp Waters Corporation ACQUITY UPLC BEH C8 Column, 130Å, 1.7 μm, 3 mm x 30 mm
Catalog No. 50791172 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The ACQUITY BEH C8 column offers less retention than C18 sorbents, enhancing speed, peak shape, and chromatographic performance. It utilizes trifunctionally bonded BEH particles, providing a wide pH range (1 to 12), superior stability, and ultra-low column bleed.
  • Less retentive C8 chemistry for faster separations and improved chromatographic performance
  • Trifunctionally bonded BEH particles provide a wide usable pH range (1 to 12), superior low pH stability, and ultra-low column bleed
  • Manufactured in a cGMP, ISO 9002 accredited facility with ultra-pure reagents; batch approved for peptide mapping
  • Ensures excellent peak asymmetry, efficiency, chemical stability, and batch-to-batch uniformity
  • Designed for extreme pH conditions and diverse small-molecule selectivity
  • Features 1.7 μm particle size, 130 Å pore size, and 3 mm x 30 mm dimensions
Catalog No. 50-791-172 Supplier Waters Corp Supplier No. 186004663
The ACQUITY BEH C8 column offers less retention than C18 sorbents, enhancing speed, peak shape, and chromatographic performance. It utilizes trifunctionally bonded BEH particles, providing a wide pH range (1 to 12), superior stability, and ultra-low column bleed.
  • Less retentive C8 chemistry for faster separations and improved chromatographic performance
  • Trifunctionally bonded BEH particles provide a wide usable pH range (1 to 12), superior low pH stability, and ultra-low column bleed
  • Manufactured in a cGMP, ISO 9002 accredited facility with ultra-pure reagents; batch approved for peptide mapping
  • Ensures excellent peak asymmetry, efficiency, chemical stability, and batch-to-batch uniformity
  • Designed for extreme pH conditions and diverse small-molecule selectivity
  • Features 1.7 μm particle size, 130 Å pore size, and 3 mm x 30 mm dimensions