Learn More
Waters Corp ACQUITY UPLC BEH C8 Column, 130Å, 1.7 µm, 3 mm X 150 mm, 3/pk
Catalog No. 50791034 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The ACQUITY BEH C8 sorbent is less retentive compared to the ACQUITY BEH C18 sorbent. A method developer may prefer the speed offered by a less retentive column, while improving peak shape and chromatographic performance. The trifunctionally bonded BEH particles offer the widest usable pH range (1-12), superior low pH stability, and ultra-low column bleed.
Catalog No. 50-791-034 Supplier Waters Corp Supplier No. 176001801
The ACQUITY BEH C8 sorbent is less retentive compared to the ACQUITY BEH C18 sorbent. A method developer may prefer the speed offered by a less retentive column, while improving peak shape and chromatographic performance. The trifunctionally bonded BEH particles offer the widest usable pH range (1-12), superior low pH stability, and ultra-low column bleed.