Learn More
Waters Corp Waters ACQUITY UPLC BEH C8 Column, 1.7 μm, 130 Å, 3 × 100 mm
Supplier:  Waters Corp 186004665
Encompass_Preferred
This ACQUITY UPLC BEH C8 Column is designed for high-performance liquid chromatography. It offers less retention than C18 sorbents, providing excellent peak shape and chromatographic performance. Manufactured in an ISO 9002 accredited facility, it ensures consistent, repeatable analysis across a wide pH range.
  • Less retentive than C18 sorbents, improving speed, peak shape, and chromatographic performance.
  • Trifunctionally bonded BEH particles provide a wide usable pH range (1–12), superior low pH stability, and ultra-low column bleed.
  • Manufactured for use with UPLCs in a cGMP, ISO 9002 accredited facility.
  • Each batch is approved for peptide mapping after chromatographic testing.
  • Precise specification ranges ensure reproducibility and consistency from batch to batch and column to column.
Catalog No. 50-791-174