Learn More
Waters Corp ACQUITY UPLC BEH C8 Method Validation Kit, 130Å, 1.7 µm, 2.1 mm X 50 mm, 3/pk
Catalog No. 50782152 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This kit consists of three ACQUITY UPLC BEH C8 columns packed with three different batches of sorbent.
Catalog No. 50-782-152 Supplier Waters Corp Supplier No. 186004046
This kit consists of three ACQUITY UPLC BEH C8 columns packed with three different batches of sorbent.