Learn More
Waters Corp ACQUITY UPLC BEH C8 Column, 1.7 μm, 3.0 x 50 mm
Catalog No. 50466027 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The ACQUITY UPLC BEH C8 Column is designed for exceptional peak asymmetry, efficiency, and chemical stability. These columns offer versatility for separating a wide range of small-molecule compounds and are robust, operating effectively under extreme pH conditions.
  • Offers exceptional peak asymmetry.
  • Provides high efficiency.
  • Features excellent chemical stability.
  • Suitable for separating a wide range of small-molecule compounds.
  • Robust design for operation under extreme pH conditions (pH range: 1–12).
  • Enables broad pH range utilization to influence retention, selectivity, and sensitivity of ionizable compounds.
Catalog No. 50-466-027 Supplier Waters Corp Supplier No. 186004693
The ACQUITY UPLC BEH C8 Column is designed for exceptional peak asymmetry, efficiency, and chemical stability. These columns offer versatility for separating a wide range of small-molecule compounds and are robust, operating effectively under extreme pH conditions.
  • Offers exceptional peak asymmetry.
  • Provides high efficiency.
  • Features excellent chemical stability.
  • Suitable for separating a wide range of small-molecule compounds.
  • Robust design for operation under extreme pH conditions (pH range: 1–12).
  • Enables broad pH range utilization to influence retention, selectivity, and sensitivity of ionizable compounds.