Learn More
Waters Corp Waters ACQUITY UPLC Glycan BEH Amide Column, 1.7 μm, 130 Å, 2.1 mm x 50 mm
Supplier:  Waters Corp 186004740
Encompass_Preferred
The Glycan BEH Amide Column is designed for efficient and repeatable separation of released glycans of biopharmaceuticals as their 2-AB derivatives. It retains polar analytes, such as carbohydrates and sugars, across a wide polarity range and is compatible with mobile phase pH 2–11. Featuring a stable, trifunctionally bonded amide phase on ethylene bridged hybrid (BEH) particle technology, it offers enhanced stability for HILIC separations. The 1.7 μm particle size enables high-speed, high-resolution analysis.
  • Optimized for glycan separation with UPLC systems.
  • Achieves retention of polar analytes across broad polarity.
  • Wide mobile phase pH compatibility (2–11).
  • Enhanced stability via BEH particle technology.
  • 1.7 μm particle size for high-speed, high-resolution analysis.
  • Extended column lifetime and increased assay robustness.
Catalog No. 50-466-060