Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters UPLC Glycan VanGuard Pre-column, BEH Amide, 1.7 μm

Catalog No. 50791198 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

The BEH Glycan Separation Technology Column is optimized for UPLC systems with fluorescence detection, separating released glycans from biopharmaceuticals as 2-aminobenzamide derivatives. This VanGuard pre-column directly connects to the analytical column's inlet, minimizing extra column volumes and leaks. It safeguards the analytical column from contaminants, enhancing performance by filtering impurities.

  • Chemistry: Amide
  • Separation mode: Hydrophilic interaction (HILIC)
  • Particle substrate: Hybrid
  • pH range: 2 to 11 pH
  • Maximum pressure: 18,000 psi (1240 Bar)
  • Particle size: 1.7 μm
  • Pore size: 130 Å
  • Endfitting type: Direct-Connect
  • Format: VanGuard Pre-column
  • System compatibility: UPLC, UHPLC
  • Inner diameter: 2.1 mm
  • Length: 5 mm
  • Molecular weight range: 1,000 to 150,000
  • Manufactured under cGMP, ISO 9001 conditions.

Catalog No. 50-791-198 Supplier Waters Corp Supplier No. 186004739
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The BEH Glycan Separation Technology Column is optimized for UPLC systems with fluorescence detection, separating released glycans from biopharmaceuticals as 2-aminobenzamide derivatives. This VanGuard pre-column directly connects to the analytical column's inlet, minimizing extra column volumes and leaks. It safeguards the analytical column from contaminants, enhancing performance by filtering impurities.

  • Chemistry: Amide
  • Separation mode: Hydrophilic interaction (HILIC)
  • Particle substrate: Hybrid
  • pH range: 2 to 11 pH
  • Maximum pressure: 18,000 psi (1240 Bar)
  • Particle size: 1.7 μm
  • Pore size: 130 Å
  • Endfitting type: Direct-Connect
  • Format: VanGuard Pre-column
  • System compatibility: UPLC, UHPLC
  • Inner diameter: 2.1 mm
  • Length: 5 mm
  • Molecular weight range: 1,000 to 150,000
  • Manufactured under cGMP, ISO 9001 conditions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.