Learn More
Waters Corp Waters ACQUITY UPLC BEH HILIC Column, 130Å, 1.7 μm, 1 mm × 100 mm
Catalog No. 50781790 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
ACQUITY BEH HILIC columns retain and separate very polar compounds using unbonded BEH substrate. Optimized for efficient, reproducible HPLC HILIC separations, they offer enhanced ruggedness and chemical stability for long column lifetimes. They utilize Hydrophilic-Interaction Chromatography (HILIC) for improved retention and orthogonal separation of polar and ionizable chemical mixtures.
  • Retains and separates very polar compounds
  • Optimized for efficient, reproducible HPLC HILIC separations
  • Enhanced ruggedness and chemical stability for long column lifetimes
  • Employs Hydrophilic-Interaction Chromatography (HILIC)
  • Offers orthogonal separation for polar and ionizable chemical mixtures
  • Achieves unparalleled efficiency, peak symmetry, and chemical stability
Catalog No. 50-781-790 Supplier Waters Corp Supplier No. 186003458
ACQUITY BEH HILIC columns retain and separate very polar compounds using unbonded BEH substrate. Optimized for efficient, reproducible HPLC HILIC separations, they offer enhanced ruggedness and chemical stability for long column lifetimes. They utilize Hydrophilic-Interaction Chromatography (HILIC) for improved retention and orthogonal separation of polar and ionizable chemical mixtures.
  • Retains and separates very polar compounds
  • Optimized for efficient, reproducible HPLC HILIC separations
  • Enhanced ruggedness and chemical stability for long column lifetimes
  • Employs Hydrophilic-Interaction Chromatography (HILIC)
  • Offers orthogonal separation for polar and ionizable chemical mixtures
  • Achieves unparalleled efficiency, peak symmetry, and chemical stability