Learn More
Waters Corp ACQUITY UPLC BEH HILIC Column, 130Å, 1.7 μm, 2.1 mm x 75 mm
Catalog No. 50466144 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
ACQUITY BEH HILIC columns contain unbonded BEH substrate, designed for the retention and separation of very polar compounds. These columns are optimized for efficient, reproducible separations under HPLC HILIC conditions. The BEH particle offers enhanced chemical stability compared to silica-based HILIC phases, leading to longer column lifetimes. They utilize Hydrophilic-Interaction Chromatography (HILIC) to improve retention of extremely polar species, providing an orthogonal separation mode for polar and ionizable chemical combinations.
  • Exceptional efficiency, peak symmetry, and chemical stability.
  • Stable across a wide pH range (1–9) in the mobile phase for very polar analytes.
  • Operating pressure up to 18,000 psi (1240 Bar).
  • Packing materials are specifically designed to ensure optimal performance.
Catalog No. 50-466-144 Supplier Waters Corp Supplier No. 186005608
ACQUITY BEH HILIC columns contain unbonded BEH substrate, designed for the retention and separation of very polar compounds. These columns are optimized for efficient, reproducible separations under HPLC HILIC conditions. The BEH particle offers enhanced chemical stability compared to silica-based HILIC phases, leading to longer column lifetimes. They utilize Hydrophilic-Interaction Chromatography (HILIC) to improve retention of extremely polar species, providing an orthogonal separation mode for polar and ionizable chemical combinations.
  • Exceptional efficiency, peak symmetry, and chemical stability.
  • Stable across a wide pH range (1–9) in the mobile phase for very polar analytes.
  • Operating pressure up to 18,000 psi (1240 Bar).
  • Packing materials are specifically designed to ensure optimal performance.