Learn More
Waters Corp Waters ACQUITY UPLC BEH HILIC Column, 130Å Pore, 1.7 μm Particle, 2.1 mm × 150 mm
Catalog No. 50780032 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
ACQUITY BEH HILIC columns are designed for the retention and separation of very polar compounds. These columns are optimized and tested for efficient and reproducible separations under HPLC HILIC conditions, offering more rugged performance than silica-based HILIC phases.
  • Contain unbonded BEH substrate
  • Designed to retain and separate very polar compounds
  • Optimized and tested to produce efficient, reproducible separations under HPLC HILIC conditions
  • More rugged than silica-based HILIC phases
  • BEH particle’s chemical stability results in long column lifetimes
  • Can operate under pressure of up to 18,000 psi (1240 bar)
Catalog No. 50-780-032 Supplier Waters Corp Supplier No. 176001094
ACQUITY BEH HILIC columns are designed for the retention and separation of very polar compounds. These columns are optimized and tested for efficient and reproducible separations under HPLC HILIC conditions, offering more rugged performance than silica-based HILIC phases.
  • Contain unbonded BEH substrate
  • Designed to retain and separate very polar compounds
  • Optimized and tested to produce efficient, reproducible separations under HPLC HILIC conditions
  • More rugged than silica-based HILIC phases
  • BEH particle’s chemical stability results in long column lifetimes
  • Can operate under pressure of up to 18,000 psi (1240 bar)